Bromothymol blue sodium salt ACS
Supplier: Thermo Scientific
ACRO403260100EA 50.3 EUR
ACRO403260100
Bromothymol blue sodium salt ACS
Bromothymol blue sodium salt
Order Now
Specification Test Results
Appearance Dark green to orange-brown Powder
Infrared spectrum Conforms
Visual transition interval from pH 6.0 (yellow) to pH 7.6 (blue)
Clarity of solution Passes test