Bromothymol blue sodium salt ACS
Supplier: Thermo Scientific
ACRO403260100EA
50.3
EUR
ACRO403260100
Bromothymol blue sodium salt ACS
Bromothymol blue sodium salt
Specification Test Results
Appearance | Dark green to orange-brown Powder |
Infrared spectrum | Conforms |
Visual transition interval | from pH 6.0 (yellow) to pH 7.6 (blue) |
Clarity of solution | Passes test |