Eriochrome Black T, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Warning
Synonyms: 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T
M-Clarity™ quality level = MQ200
858390-100G 858390-1KG 858390-500G
858390-100GEA 64.1 EUR
858390-100G SIAL858390-1KG SIAL858390-500G
Eriochrome Black T, Sigma-Aldrich®
Eriochrome Black T
Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9 III
Merck Index: 13,03702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a54fcee283659c6154da4eddffaef1718d046925&vid=8f1128967a01a32802f875d8c8d77de19679a735&prid=web&clid=SAPDAM