3-Thiopheneboronic acid, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Thienylboronic acid, Thiophene-3-boronic acid
M-Clarity™ quality level = MQ100
436844-1G 436844-5G
SIAL436844-1GEA 34.4 EUR
SIAL436844-1G SIAL436844-5G
3-Thiopheneboronic acid, Sigma-Aldrich®
3-Thiopheneboronic acid
Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00151851
CAS Number: 6165-69-1