Sodium alginate, very low viscosity
Supplier: Thermo Scientific
Synonyms: Alginic acid sodium salt, Algin
A18565.22 A18565.0E A18565.36
A18565.22EA 46.7 EUR
A18565.22 A18565.0E A18565.36
Sodium alginate, very low viscosity
Sodium alginate
CAS No.: 9005-38-3 Merck Reference: 14,241 MDL No.: MFCD00081310
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241