Lead(II) perchlorate hydrate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
205311-250G
205311-50G
SIAL205311-250GEA
129
EUR
SIAL205311-250G
SIAL205311-50G
Lead(II) perchlorate hydrate, Sigma-Aldrich®
Lead (II) perchlorate hydrate
Formula:
Cl₂O₈Pb·H₂O MW: 424.12 g/mol |
CAS Number:
207500-00-3 EINECS: 237-125-7 UN: 1470 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...