Hexafluorosilicic acid 35% (w/w) in aqueous solution
Supplier: Thermo Scientific
L14829.22EA 26.3 EUR
L14829.22 L14829.36 L14829.0E L14829.PG
Hexafluorosilicic acid 35% (w/w) in aqueous solution
Hexafluorosilicic acid
Formula: H₂F₆Si
MW: 144,09 g/mol
Boiling Pt: 108…109 °C
Density: 1,320
Storage Temperature: Ambient
MDL Number: MFCD00036289
CAS Number: 16961-83-4
UN: 1778
ADR: 8,II
Merck Index: 14,4182