Potassium dihydrogen phosphate 1 mol/l in aqueous solution, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
P8709-1L P8709-4L
SIALP8709-1LEA 101 EUR
SIALP8709-1L SIALP8709-4L
Potassium dihydrogen phosphate 1 mol/l in aqueous solution, Sigma-Aldrich®
Potassium dihydrogen phosphate
Formula: KH₂PO₄
Storage Temperature: Ambient
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4