Pyridinium dichromate 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: PDC
200880500 200882500 200880010
ACRO200880500EA 33.9 EUR
ACRO200880500 ACRO200882500 ACRO200880010
Pyridinium dichromate 98%
Pyridinium dichromate
Formula: C₁₀N₁₂Cr₂N₂O₇
MW: 532.19 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00013105
CAS Number: 20039-37-6
EINECS: 243-478-8
UN: 3085
ADR: 5.1 II
Order Now
Specification Test Results
Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %