(±)-trans-1,2-Diaminocyclohexane, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine
M-Clarity™ quality level = MQ200
270016-10ML 270016-250ML 270016-50ML
SIAL270016-10MLEA 35.7 EUR
SIAL270016-10ML SIAL270016-250ML SIAL270016-50ML
(±)-trans-1,2-Diaminocyclohexane, Sigma-Aldrich®
(±)-trans-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 14.8 °C
Density: 0.951 g/cm³ (20 °C)
MDL Number: MFCD00063747
CAS Number: 1121-22-8