Anti-ACA125 177 Lu

Supplier: ProSci
PRSI10-136EA 627 EUR
PRSI10-136
Anti-ACA125 177 Lu
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR