Anti-MEDI-522 Recombinant Antibody

Supplier: ProSci
PRSI10-260EA 627 EUR
PRSI10-260
Anti-MEDI-522 Recombinant Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR