Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PacBlue)

Supplier: AAT BIOQUEST
100211J0 100211J1
AATB100211J0EA 371 EUR
AATB100211J0 AATB100211J1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (PacBlue)
Antibodies
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Upozornení: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: PacBlue
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR