Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 568)
Supplier: AAT BIOQUEST
104410B0 104410B1
AATB104410B0EA 371 EUR
AATB104410B0 AATB104410B1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (iFluor® 568)
Antibodies
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Upozornení: For research use only.
Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: iFluor® 568
Epitope:
Host: Rat
Isotype:
Reactivity: Human