Anti-ESAM Rabbit Ployclonal Antibody (AbBy Fluor® 647)

Supplier: Bioss
BOSSBS-5838R-BF647EA 699 EUR
BOSSBS-5838R-BF647
Anti-ESAM Rabbit Ployclonal Antibody (AbBy Fluor® 647)
Antibodies
Type: Primary
Antigen: ESAM
Clonality: Ployclonal
Clone:
Conjugation: AbBy Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR