Septum Stopper, Sleeve Type
Supplier: Chemglass
VWRU89029-091EA 55.97 EUR
VWRU89029-091
Septum Stopper, Sleeve Type
Stoppers Rubber Stoppers
Sleeve type stopper with hollow plugs and sleeve extensions.
  • Available in red or white rubber
The sleeve extensions fold down over the neck of a vessel and tubing. The diaphragm can be punctured with a syringe needle, re-sealing after the needle is withdrawn.