Industrial Scoops
Supplier: Sampling Systems
A643-500 A643-100 A643-200 A643-300
SAMPA643-500EA 59.3 EUR
SAMPA643-500 SAMPA643-100 SAMPA643-200 SAMPA643-300
Industrial Scoops
Scoops
Industrial scoops are cost-effective devices for general-purpose use.
  • Low-cost for general-purpose use
  • Made from stainless steel
  • Various sizes available
  • Available from stock
  • Fast delivery
Industrial scoops provide a low-cost solution for all types of general-purpose work. Available in different sizes, they are ideal for working with chemicals and most foodstuffs.