Methyl-2-chloro-4-nitrobenzoate
Supplier: Thermo Scientific
Synonyms: 2-Chloro-4-nitrobenzoic acid methyl ester, Methyl 2-chloro-4-nitrobenzoate
KM08842.1 KM08842.10 KM08842.50
MAYBKM08842.1EA 0 EUR
MAYBKM08842.1 MAYBKM08842.10 MAYBKM08842.50
Methyl-2-chloro-4-nitrobenzoate
Methyl 2-chloro-4-nitrobenzoate
Formula: C₈H₆ClNO₄
MDL Number: MFCD00017013
CAS Number: 13324-11-3
EINECS: 236-362-3