Methyl-4-chloro-2-nitrobenzoate
Supplier: Thermo Scientific
Synonyms:
4-Chloro-2-nitrobenzoic acid methyl ester, Methyl 4-chloro-2-nitrobenzoate
SB00722.1
SB00722.10
SB00722.50
MAYBSB00722.1EA
0
EUR
MAYBSB00722.1
MAYBSB00722.10
MAYBSB00722.50
Methyl-4-chloro-2-nitrobenzoate
Methyl 4-chloro-2-nitrobenzoate
Formula:
C₈H₆ClNO₄ MW: 215.59 g/mol |
MDL Number:
MFCD00007213 CAS Number: 42087-80-9 EINECS: 255-654-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...