Eosin Y (yellowish), high purity biological stain
Supplier: Thermo Scientific
Warning
Synonyms: Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
152880250 152881000 152885000
ACRO152880250EA 29.2 EUR
ACRO152880250 ACRO152881000 ACRO152885000
Eosin Y (yellowish), high purity biological stain
Eosin Y (yellowish)
Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: >300 °C (dec.)
Density: 1.018 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634
Order Now
Specification Test Results
Appearance Red to brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤10 %
Dye content ≥90 % (on dry substance)