Wood's alloy (lipowitz's alloy), sticks, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Wood's metal, Bismuth Lead Cadmium Tin alloy (50:25:12.5:12.5 w%)
M-Clarity™ quality level = MQ200
244104-100G 244104-500G
SIAL244104-100GEA 98.6 EUR
SIAL244104-100G SIAL244104-500G
Wood's alloy (lipowitz's alloy), sticks, Sigma-Aldrich®
Wood's alloy (Lipowitz's alloy)
Melting Pt: 75 °C
Density: 9.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198175
CAS Number: 76093-98-6
UN: 3288
ADR: 6.1,II