Wood's alloy (lipowitz's alloy)
Supplier: Roth Carl
Synonyms: Wood's metal, Bismuth Lead Cadmium Tin alloy (50:25:12.5:12.5 w%)
1980.1 1980.2
ROTH1980.1EA 58.9 EUR
ROTH1980.1 ROTH1980.2
Wood's alloy (lipowitz's alloy)
Wood's alloy (Lipowitz's alloy)
Melting Pt: 75 °C
Density: 9.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198175
CAS Number: 76093-98-6