Methyl docosanoate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methyl behenate, Docosanoic acid methyl ester, Behenic acid methyl ester

M-Clarity™ quality level = MQ200

B3271-1G B3271-5G
SIALB3271-1GEA 253 EUR
SIALB3271-1G SIALB3271-5G
Methyl docosanoate, Sigma-Aldrich®
Methyl docosanoate
Formula: C₂₃H₄₆O₂
MW: 354.62 g/mol
MDL Number: MFCD00009347
CAS Number: 929-77-1
EINECS: 213-207-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR