F(ab')2 Anti-IgG gamma Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: SeraCare
KPLIKPL-2141002EA 720 EUR
KPLIKPL-2141002
F(ab')2 Anti-IgG gamma Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Affinity-purified polyclonal antibody, F(ab')2 fragment, to human IgG heavy chains (gamma), made in goat, and labeled with horseradish peroxidase. Product is in lyophilised form. Each lot is tested to assure specificity and lot-to-lot consistency using an in-house ELISA assay.

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR