Safety Shoes, Lace-Up, Low, Tegmen IV
Supplier: Basan
93875-42 93875-46 93875-40
BSNC93875-42EA 204 EUR
BSNC93875-42 ENGE93875-46 ENGE93875-40
Safety Shoes, Lace-Up, Low, Tegmen IV
Shoes and Boots
The air-permeable knitting technology not only makes the Tegmen IV low an eye-catcher, the outer material made of tough polyamide fibres in an almost seamless finish also ensures that the shoes are incredibly durable.
  • Pleasantly light and breathable in a trendy knitted look
  • Proven knitting technology, almost seamless outer material made of polyamide fibres
  • BOA® Fit System for a micro-adjustable precision fit
  • Pleasant textile inner lining with free of leather
  • Removable, full-surface anatomically shaped insole
  • Also ideal for insole wearers
  • Anti-slip and comfortable absorbent rubber/phylon sole conforms to SRC is anti-static, fuel and heat resistant up to approx. 70 °C
The easy-to-use rotary fastener means that the shoes can be put on and taken off very quickly; the ultra-light aluminium cap ensures that these sporty shoes are among the lightest available in the safety shoes sector.
Certifikace: EN ISO 20345:2011 S1 with an aluminium toe cap.