Eosin Y (yellowish) 1% ethanolic aqueous solution (CE-IVD) for clinical diagnosis

Supplier: PanReac AppliChem

APPC251301.1211EA 0 EUR
APPC251301.1211
Eosin Y (yellowish) 1% ethanolic aqueous solution (CE-IVD) for clinical diagnosis
Eosin Y (yellowish)

Eosin yellowish hydroalcoholic solution 1% for clinical diagnostics solution for Hematoxylin-Eosin staining, in human and gynaecological samples.


Composition:
Yellowish eosin: 1 g
Absolute ethanol: 10 ml
Water: 90 ml


Maximum limit of impurities suitability for hematoxylin - eosin.


Certifikace: CE-IVD, Class A in vitro diagnostic medical device in accordance with Regulation (EU) 2017/746.

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691,86 g/mol
Density: ~0,997 kg/l
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR