Coleman® C500 Performance Gas Cartridge
Supplier: Camping Gaz
Coleman®
CAMP2182837EA 8.7 EUR
CAMP2182837
Coleman® C500 Performance Gas Cartridge
Heaters Gas Cartridges
Coleman® C500 Performance gas cartridge with safety valve.
  • Filled with 70% butane and 30% propane gas
  • Capacity: 440 g
  • Dimensions: 11Ø×14H cm
  • Weight: 598 g
Ideal for sporadic, moderate use, this gas cartridge is equipped with a high security, self-sealing valve. The cartridge can be disconnected and reconnected even when it is not empty.