Anti-CD4 antigen Goat Monoclonal Antibody [clone: SK3] (mFluor™ Blue 585)
Supplier: AAT BIOQUEST
100421Z0 100421Z1
AATB100421Z0EA 341 EUR
AATB100421Z0 AATB100421Z1
Anti-CD4 antigen Goat Monoclonal Antibody [clone: SK3] (mFluor™ Blue 585)
Antibodies
SK3 is an anti-human monoclonal antibody that recognizes the CD4 antigen.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
It is compatible with the 488 nm laser and 586/15 nm bandpass filter (for example, as in the BD FACSCelesta™).
Upozornení: Research use only.
Type: Secondary
Antigen: CD4 antigen
Clonality: Monoclonal
Clone: SK3
Conjugation: mFluor™ Blue 585
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human