iFluor® 540 succinimidyl ester
Supplier: AAT BIOQUEST INC
AATB1426EA 297 EUR
AATB1426
iFluor® 540 succinimidyl ester
iFluor® 540 succinimidyl ester
iFluor® 540 succinimidyl ester is the most convenient reactive form of iFluor® 540 dyes that can be readily used for labeling proteins such as antibodies.
  • Stable for 12 months
Quality tested in-house
Upozornení: For Research Use Only