ReadiUse™ 1 Kb Plus DNA Ladder
Supplier: AAT BIOQUEST
60051
60050
AATB60051EA
283
EUR
AATB60051
AATB60050
ReadiUse™ 1 Kb Plus DNA Ladder
Electrophoresis Reagents
Molecular Weight Markers
ReadiUse™ 1 kb Plus DNA ladder is a ready-to-use DNA ladder, which is pre-mixed with loading dye for direct gel loading.
- Stable for 12 months
Quality tested in-house.
Upozornení: For research use only.