Captair® 632A Midcap Ductless Filtering Storage Cabinet
Supplier: Erlab
Captair®
ERLAAB072AA0000003EA 1233 EUR
ERLAAB072AA0000003
Captair® 632A Midcap Ductless Filtering Storage Cabinet
Furniture Storage Cabinets
This chemical storage cabinets provide outstanding filtration quality which ensure users and laboratory technicians protection against chemical hazards.
  • The safest storage cabinets are also the least expensive
  • Designed to keep users free from chemical inhalations
  • Filtering storage cabinets minimize the risk posed by fumes, vapors, and chemicals
  • A level of protection above even the most stringent standards
  • Equipment that is easy to install and simple to use
  • Filtering storage cabinets connect to your smartphone and send you valuable information in real time
  • Increased safety from a door sensor which warns the user if they are open for a prolonged period of time
Their technology adapts to the type of product being stored. Users are given the best possible view of the stored products through their transparent doors and are alerted in the event of prolonged opening.
Air flow: 10 m³/h
Voltage: 110 - 230 V/50 - 60 Hz
Power consumption: 16 W
Structure: Corrosion resistant electro-galvanized steel coated with anti-acid polymer
Filtration module: Polypropylene
Particulate filtration for powders*: HEPA H13: 99,95 % efficiency filtration of particles over 0,1 μm in size
Lock: key lock
Certifikace: According to the AFNOR NF X 15-211:2009 standard : France, EN 1822 : 1998 (HEPA H14 Filters).
Delivery information: Supplied with two retention trays and two sliding shelves, and two absorbant mats.