VWR® Miscellaneous Dissolution Accessories
Supplier: Avantor
VWR®
VWRU75856-092EA 427.75 EUR
VWRU75856-092
VWR® Miscellaneous Dissolution Accessories
Dissolution Apparatus
Everclear water bath preservative is a crystalline formula offered in premeasured vials for use with dissolution water baths. Use with DEIONIZED water.
The following products are compatible with Agilent/VanKel equipment: APP7 accessories, rotating bottle accessories, Auto-sampler accessories and Paper Rolls.
PTFE suspension cups can be used with most dissolution baths.
A single dose will protect a bath for up to 8 weeks.