(4-Nitrophenyl)-β-D-glucopyranoside, Sigma-Aldrich®

Supplier: Merck

Synonyms: p-Nitrophenyl-β-D-Glucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

M-Clarity™ quality level = MQ300

N7006-1G N7006-25G N7006-500MG N7006-5G
SIALN7006-1GEA 114 EUR
SIALN7006-1G SIALN7006-25G SIALN7006-500MG SIALN7006-5G
(4-Nitrophenyl)-β-D-glucopyranoside, Sigma-Aldrich®
(4-Nitrophenyl) β-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR