EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide), Sigma-Aldrich®
Supplier: Merck
Synonyms: EDAC, WSC, EDC, N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, N1-((Ethylimino)methylene)-N3,N3-dimethylpropane-1,3-diamine
39391-10ML 39391-50ML
SIAL39391-10MLEA 152 EUR
SIAL39391-10ML SIAL39391-50ML
EDC (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide), Sigma-Aldrich®
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide
Formula: C₈H₁₇N₃
MW: 155.24 g/mol
Boiling Pt: 43…45 °C (0.3 mmHg)
Melting Pt: 114…115 °C
Density: 0.885 g/cm³ (20 °C)
MDL Number: MFCD00044916
CAS Number: 1892-57-5
EINECS: 217-579-2