EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
M-Clarity™ quality level = MQ200
E6383-1G
E6383-5G
SIALE6383-1GEA
58.4
EUR
SIALE6383-1G
SIALE6383-5G
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride), Sigma-Aldrich®
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride
Formula:
C₈H₁₇N₃·HCl MW: 191.7 g/mol Melting Pt: 112…113 °C Storage Temperature: Freezer |
MDL Number:
MFCD00012503 CAS Number: 25952-53-8 EINECS: 247-361-2 |