4-Amino-N,N-diethylaniline, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N,N-Diethyl-1,4-phenylenediamine, N,N-Diethyl-p-phenylenediamine, N,N-Diethylbenzene-1,4-diamine, N,N-Diethylbenzene-p-diamine, N-1,N-1-Diethyl-1,4-benzenediamine
M-Clarity™ quality level = MQ100
261513-100G 261513-25G
SIAL261513-100GEA 150 EUR
SIAL261513-100G SIAL261513-25G
4-Amino-N,N-diethylaniline, Sigma-Aldrich®
4-Amino-N,N-diethylaniline
Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 260
Melting Pt: 23
Density: 0.988 g/cm³ (20 °C)
Flash Pt: 139
MDL Number: MFCD00007861
CAS Number: 93-05-0
EINECS: 202-214-1