2-Naphthoyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Naphthoyl Chloride
M-Clarity™ quality level = MQ100
250260-10G 250260-50G
SIAL250260-10GEA 109 EUR
SIAL250260-10G SIAL250260-50G
2-Naphthoyl chloride, Sigma-Aldrich®
2-Naphthoyl chloride
Formula: C₁₁H₇ClO
MW: 190.63 g/mol
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 49…52 °C
MDL Number: MFCD00004093
CAS Number: 2243-83-6
EINECS: 218-822-5
UN: 3261
ADR: 8,II