Nitro blue tetrazolium chloride (NBT)
Supplier: Roth Carl
Warning
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
4421.1 4421.2 4421.3
ROTH4421.1EA 23.1 EUR
ROTH4421.1 ROTH4421.2 ROTH4421.3
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)
Formula: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817.65 g/mol
Melting Pt: 200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012159
CAS Number: 298-83-9
EINECS: 206-067-4