Nitro blue tetrazolium chloride (NBT)
Supplier: Biotium
|
Warning
|
Synonyms:
5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
10008-1
10008-2
10008
10008-1EA
231
EUR
10008-1
10008-2
10008.
Nitro blue tetrazolium chloride (NBT)
Nitro blue tetrazolium chloride (NBT)
Formula:
C₄₀H₃₀Cl₂N₁₀O₆ MW: 817.65 g/mol Melting Pt: 200 Storage Temperature: Refrigerator |
MDL Number:
MFCD00012159 CAS Number: 298-83-9 EINECS: 206-067-4 |