X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
B4252-100MG B4252-1G B4252-250MG B4252-2G B4252-500MG B4252-50MG B4252-5G
SIALB4252-100MGEA 105 EUR
SIALB4252-100MG SIALB4252-1G SIALB4252-250MG SIALB4252-2G SIALB4252-500MG SIALB4252-50MG SIALB4252-5G
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside), Sigma-Aldrich®
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
Melting Pt: 237…239 °C
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8