X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Supplier: G Bioscienes
RC-113EA 89.8 EUR
RC-113
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Formula: C₁₄H₁₅BrClNO₆
MW: 408,63 g/mol
Melting Pt: 237…239 °C
MDL Number: MFCD00005666
CAS Number: 7240-90-6