Dextran, M.W. ~ 40,000, BioChemika®, Sigma-Aldrich®
Supplier: Merck
31389-100G 31389-25G 31389-500G
SIAL31389-100GEA 346 EUR
SIAL31389-100G SIAL31389-25G SIAL31389-500G
Dextran, M.W. ~ 40,000, BioChemika®, Sigma-Aldrich®
Dextran
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5