Dextran, M.W. ~ 6,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Supplier: Merck
31388-100G 31388-25G 31388-500G
SIAL31388-100GEA 443 EUR
SIAL31388-100G SIAL31388-25G SIAL31388-500G
Dextran, M.W. ~ 6,000 (from Leuconostoc sp.), BioChemika®, Sigma-Aldrich®
Dextran
Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5