Dicyclohexylcarbodiimide, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D80002-100G D80002-25G D80002-1KG
SIALD80002-100GEA 27.3 EUR
SIALD80002-100G SIALD80002-25G SIALD80002-1KG
Dicyclohexylcarbodiimide, Sigma-Aldrich®
Dicyclohexylcarbodiimide
Formula: C₁₃H₂₂N₂
Boiling Pt: 148…152 °C (15 hPa)
Melting Pt: 35…36 °C
Density: 0.95 g/cm³ (40 °C)
Flash Pt: 113 °C
MDL Number: MFCD00011659
CAS Number: 538-75-0
EINECS: 208-704-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,03123