Silica gel, granules -6+12 mesh desiccant
Supplier: Thermo Fisher Scientific
44387.30 44387.A1 44387.A7
44387.30EA 37.6 EUR
44387.30 44387.A1 44387.A7
Silica gel, granules -6+12 mesh desiccant
Silica gel
CAS No.: 7631-86-9 Sensitivity: Hygroscopic Odour: Odourless MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
REACH: 01-2119379499-16