Silica gel, Pore size: 120 Å 99,99, spherical 7.5 µm APS Chromatography_prep
Supplier: Thermo Fisher Scientific
Warning
45816.14 45816.06
45816.14EA 242 EUR
45816.14 45816.06
Silica gel, Pore size: 120 Å 99,99, spherical 7.5 µm APS Chromatography_prep
Silica gel
Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 335m2/g, P.V. 1.00cc/g MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4