Silica gel, Pore size: 300 Å, spherical
Supplier: Thermo Fisher Scientific
Warning
44101.09 44101.18
44101.09EA 212 EUR
44101.09 44101.18
Silica gel, Pore size: 300 Å, spherical
Silica gel
CAS No.: 63231-67-4 Physical Form: 20 Micron APS, S.A. 100m2/g, P.V. 0.9cc/g MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4