Silica gel, Pore size: 60 Å ≥99.99%, spherical 10 µm APS for preparative chromatography
Supplier: Thermo Fisher Scientific
Warning
45819.06 45819.14
45819.06EA 56.6 EUR
45819.06 45819.14
Silica gel, Pore size: 60 Å ≥99.99%, spherical 10 µm APS for preparative chromatography
Silica gel
Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 650m2/g, P.V. 1.00cc/g MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4