Silica gel, Pore size: 70 Å 99,99, spherical 5 µm APS HPLC
Supplier: Thermo Fisher Scientific
Warning
45809.06 45809.14
45809.06EA 145 EUR
45809.06 45809.14
Silica gel, Pore size: 70 Å 99,99, spherical 5 µm APS HPLC
Silica gel
Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 500m2/g, P.V. 0.95cc/g Odour: Odourless pH: 5.5-8 MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4