Florisil® 100-200 mesh for chromatography
Supplier: Thermo Scientific
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
B21870.0B B21870.30
B21870.0BEA 388 EUR
B21870.0B B21870.30
Florisil® 100-200 mesh for chromatography
Magnesium silicate
CAS No.: 1343-88-0 MDL No.: MFCD00078249
Formula: MgO₃Si
MW: 100.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1