Florisil® 60-100 mesh for chromatography
Supplier: Thermo Fisher Scientific
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
205450010 205452500 205455000 205450050
ACRO205450010EA 355 EUR
ACRO205450010 ACRO205452500 ACRO205455000 ACRO205450050
Florisil® 60-100 mesh for chromatography
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now
Specification Test Results
Appearance White to off-white Powder
Particle size 6.9 % (140 mesh) typical